COM448685180 Combi-Blocks, Inc.

2-Chloro-5-(2,4-dichlorophenyl)phenol

  • MDL number MFCD18315721
  • Cas Number : 1227375-87-2
  • Molecular Weight : 273.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4818
Availability details for
Quantity
  • Availability
  • Add to Cart