COM448685184 Combi-Blocks, Inc.

3-Bromo-5-(4-hydroxyphenyl)phenol

  • MDL number MFCD18316029
  • Cas Number : 1261956-89-1
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-5131

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-5131
Availability details for
Quantity
  • Availability
  • Add to Cart