COM448685185 Combi-Blocks, Inc.

3-Bromo-5-(4-carboxyphenyl)phenol

  • MDL number MFCD18316077
  • Cas Number : 1261991-91-6
  • Molecular Weight : 293.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-5179

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-5179
Availability details for
Quantity
  • Availability
  • Add to Cart