COM448685190 Combi-Blocks, Inc.

5-(3,4-Difluorophenyl)-2-hydroxypyrimidine

  • MDL number MFCD11876836
  • Cas Number : 1111108-31-6
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-5695

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-5695
Availability details for
Quantity
  • Availability
  • Add to Cart