COM448685205 Combi-Blocks, Inc.

6-(Thiophen-2-yl)nicotinic acid

  • MDL number MFCD06410384
  • Cas Number : 179408-54-9
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-5959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-5959
Availability details for
Quantity
  • Availability
  • Add to Cart