COM448685212 Combi-Blocks, Inc.

6-(2-Methylphenyl)picolinic acid

  • MDL number MFCD06410063
  • Cas Number : 887983-38-2
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-5997

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-5997
Availability details for
Quantity
  • Availability
  • Add to Cart