COM448685215 Combi-Blocks, Inc.

3-(3-Methylphenyl)picolinic acid

  • MDL number MFCD16308583
  • Cas Number : 1225956-74-0
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6011
Availability details for
Quantity
  • Availability
  • Add to Cart