COM448685220 Combi-Blocks, Inc.

5-(4-Methylphenyl)picolinic acid

  • MDL number MFCD16308584
  • Cas Number : 1225956-67-1
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6030
Availability details for
Quantity
  • Availability
  • Add to Cart