COM448685221 Combi-Blocks, Inc.

6-(4-Methylphenyl)picolinic acid

  • MDL number MFCD06410065
  • Cas Number : 86696-72-2
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6031
Availability details for
Quantity
  • Availability
  • Add to Cart