COM448685228 Combi-Blocks, Inc.

3-(4-Hydroxyphenyl)picolinic acid

  • MDL number MFCD18085819
  • Cas Number : 1258622-77-3
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6096
Availability details for
Quantity
  • Availability
  • Add to Cart