COM448685242 Combi-Blocks, Inc.

6-(4-Cyanophenyl)picolinic acid

  • MDL number MFCD18316810
  • Cas Number : 1261929-53-6
  • Molecular Weight : 224.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6201

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6201
Availability details for
Quantity
  • Availability
  • Add to Cart