COM448685246 Combi-Blocks, Inc.

5-(4-Ethylphenyl)nicotinic acid

  • MDL number MFCD16200865
  • Cas Number : 1048308-04-8
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6298
Availability details for
Quantity
  • Availability
  • Add to Cart