COM448685248 Combi-Blocks, Inc.

4-(2,4-Dimethylphenyl)picolinic acid

  • MDL number MFCD18316887
  • Cas Number : 1261967-40-1
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6318

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6318
Availability details for
Quantity
  • Availability
  • Add to Cart