COM448685250 Combi-Blocks, Inc.

6-(2,4-Dimethylphenyl)picolinic acid

  • MDL number MFCD16314268
  • Cas Number : 1225538-24-8
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6320

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6320
Availability details for
Quantity
  • Availability
  • Add to Cart