COM448685257 Combi-Blocks, Inc.

2-(4-Methoxyphenyl)isonicotinic acid

  • MDL number MFCD09909443
  • Cas Number : 935861-30-6
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6431
Availability details for
Quantity
  • Availability
  • Add to Cart