COM448685262 Combi-Blocks, Inc.

6-(5-Fluoro-2-methylphenyl)picolinic acid

  • MDL number MFCD18317058
  • Cas Number : 1261896-76-7
  • Molecular Weight : 231.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6541

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6541
Availability details for
Quantity
  • Availability
  • Add to Cart