COM448685265 Combi-Blocks, Inc.

5-(3-Chlorophenyl)nicotinic acid

  • MDL number MFCD06410265
  • Cas Number : 375853-95-5
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6672

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6672
Availability details for
Quantity
  • Availability
  • Add to Cart