COM448685268 Combi-Blocks, Inc.

3-(4-Chlorophenyl)isonicotinic acid

  • MDL number MFCD18085888
  • Cas Number : 883107-39-9
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6687
Availability details for
Quantity
  • Availability
  • Add to Cart