COM448685275 Combi-Blocks, Inc.

5-(4-Chlorophenyl)-2-methoxynicotinic acid

  • MDL number MFCD18317183
  • Cas Number : 1261905-29-6
  • Molecular Weight : 263.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6700
Availability details for
Quantity
  • Availability
  • Add to Cart