COM448685277 Combi-Blocks, Inc.

5-(2,4-Difluorophenyl)nicotinic acid

  • MDL number MFCD06410287
  • Cas Number : 887973-46-8
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6706

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6706
Availability details for
Quantity
  • Availability
  • Add to Cart