COM448685282 Combi-Blocks, Inc.

6-(3,5-Difluorophenyl)picolinic acid

  • MDL number MFCD11847143
  • Cas Number : 1226098-50-5
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6745
Availability details for
Quantity
  • Availability
  • Add to Cart