COM448685287 Combi-Blocks, Inc.

5-(3-Carboxyphenyl)nicotinic acid

  • MDL number MFCD18317323
  • Cas Number : 1261976-89-9
  • Molecular Weight : 243.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6893

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6893
Availability details for
Quantity
  • Availability
  • Add to Cart