COM448685288 Combi-Blocks, Inc.

5-(4-Carboxyphenyl)nicotinic acid

  • MDL number MFCD18317340
  • Cas Number : 597565-52-1
  • Molecular Weight : 243.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6910

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6910
Availability details for
Quantity
  • Availability
  • Add to Cart