COM448685290 Combi-Blocks, Inc.

5-(3-Nitrophenyl)nicotinic acid

  • MDL number MFCD06410280
  • Cas Number : 898907-67-0
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7012

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7012
Availability details for
Quantity
  • Availability
  • Add to Cart