COM448685293 Combi-Blocks, Inc.

5-(4-Fluoro-3-methoxyphenyl)nicotinic acid

  • MDL number MFCD18317538
  • Cas Number : 1261934-09-1
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7148
Availability details for
Quantity
  • Availability
  • Add to Cart