COM448685296 Combi-Blocks, Inc.

5-(4-Chloro-3-methylphenyl)-6-hydroxynicotinic acid

  • MDL number MFCD18317651
  • Cas Number : 1262006-39-2
  • Molecular Weight : 263.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7277

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7277
Availability details for
Quantity
  • Availability
  • Add to Cart