COM448685300 Combi-Blocks, Inc.

5-(4-t-Butylphenyl)picolinic acid

  • MDL number MFCD16329814
  • Cas Number : 1226273-30-8
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7407
Availability details for
Quantity
  • Availability
  • Add to Cart