COM448685303 Combi-Blocks, Inc.

2-Methoxy-5-(4-t-butylphenyl)nicotinic acid

  • MDL number MFCD18317771
  • Cas Number : 1261994-19-7
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7414

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7414
Availability details for
Quantity
  • Availability
  • Add to Cart