COM448685308 Combi-Blocks, Inc.

5-(3-Chloro-4-methoxyphenyl)nicotinic acid

  • MDL number MFCD16200869
  • Cas Number : 1261960-17-1
  • Molecular Weight : 263.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7709

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7709
Availability details for
Quantity
  • Availability
  • Add to Cart