COM448685311 Combi-Blocks, Inc.

5-(3-Trifluoromethylphenyl)nicotinic acid

  • MDL number MFCD06410275
  • Cas Number : 893740-46-0
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7777
Availability details for
Quantity
  • Availability
  • Add to Cart