COM448685312 Combi-Blocks, Inc.

5-(3-Trifluoromethylphenyl)picolinic acid

  • MDL number MFCD12406460
  • Cas Number : 87789-84-2
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7781
Availability details for
Quantity
  • Availability
  • Add to Cart