COM448685314 Combi-Blocks, Inc.

5-(4-Trifluoromethylphenyl)picolinic acid

  • MDL number MFCD12406459
  • Cas Number : 762287-48-9
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7798
Availability details for
Quantity
  • Availability
  • Add to Cart