COM448685317 Combi-Blocks, Inc.

5-(3,4-Dichlorophenyl)nicotinic acid

  • MDL number MFCD09042512
  • Cas Number : 926255-89-2
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7862
Availability details for
Quantity
  • Availability
  • Add to Cart