COM448685318 Combi-Blocks, Inc.

5-(3,5-Dichlorophenyl)nicotinic acid

  • MDL number MFCD06410288
  • Cas Number : 887973-53-7
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-7879
Availability details for
Quantity
  • Availability
  • Add to Cart