COM448685324 Combi-Blocks, Inc.

5-(4-Methylsulfonylphenyl)nicotinic acid

  • MDL number MFCD06802759
  • Cas Number : 893740-53-9
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8083

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8083
Availability details for
Quantity
  • Availability
  • Add to Cart