COM448685325 Combi-Blocks, Inc.

2-(3-Carboxy-4-chlorophenyl)nicotinic acid

  • MDL number MFCD18318332
  • Cas Number : 1261947-93-6
  • Molecular Weight : 277.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8116

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8116
Availability details for
Quantity
  • Availability
  • Add to Cart