COM448685330 Combi-Blocks, Inc.

4-(3-Benzyloxyphenyl)picolinic acid

  • MDL number MFCD18085915
  • Cas Number : 1258625-79-4
  • Molecular Weight : 305.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8409

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8409
Availability details for
Quantity
  • Availability
  • Add to Cart