COM448685332 Combi-Blocks, Inc.

5-(4-Benzyloxyphenyl)nicotinic acid

  • MDL number MFCD06802460
  • Cas Number : 893739-18-9
  • Molecular Weight : 305.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8423

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8423
Availability details for
Quantity
  • Availability
  • Add to Cart