COM448685333 Combi-Blocks, Inc.

6-(4-Benzyloxyphenyl)nicotinic acid

  • MDL number MFCD18318602
  • Cas Number : 1261900-35-9
  • Molecular Weight : 305.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8424

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8424
Availability details for
Quantity
  • Availability
  • Add to Cart