COM448685334 Combi-Blocks, Inc.

3-(4-Benzyloxyphenyl)picolinic acid

  • MDL number MFCD18318603
  • Cas Number : 1261892-48-1
  • Molecular Weight : 305.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8425
Availability details for
Quantity
  • Availability
  • Add to Cart