COM448685340 Combi-Blocks, Inc.

6-Methyl-2-phenylbenzoic acid

  • MDL number MFCD14702119
  • Cas Number : 92254-02-9
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8641

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8641
Availability details for
Quantity
  • Availability
  • Add to Cart