COM448685341 Combi-Blocks, Inc.

3-Methyl-4-phenylbenzoic acid

  • MDL number MFCD11554176
  • Cas Number : 178313-67-2
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8642

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8642
Availability details for
Quantity
  • Availability
  • Add to Cart