COM448685344 Combi-Blocks, Inc.

3-Amino-5-phenylbenzoic acid

  • MDL number MFCD18318819
  • Cas Number : 129192-15-0
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8645

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8645
Availability details for
Quantity
  • Availability
  • Add to Cart