COM448685345 Combi-Blocks, Inc.

3-Hydroxy-4-phenylbenzoic acid

  • MDL number MFCD12973936
  • Cas Number : 106593-48-0
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8646

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8646
Availability details for
Quantity
  • Availability
  • Add to Cart