COM448685346 Combi-Blocks, Inc.

5-Hydroxy-3-phenylbenzoic acid

  • MDL number MFCD18086680
  • Cas Number : 35489-88-4
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8647

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8647
Availability details for
Quantity
  • Availability
  • Add to Cart