COM448685350 Combi-Blocks, Inc.

4-Fluoro-3-phenylbenzoic acid

  • MDL number MFCD11577106
  • Cas Number : 1082484-37-4
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8651

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8651
Availability details for
Quantity
  • Availability
  • Add to Cart