COM448685352 Combi-Blocks, Inc.

2-Fluoro-5-phenylbenzoic acid

  • MDL number MFCD09042321
  • Cas Number : 146328-84-9
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8654
Availability details for
Quantity
  • Availability
  • Add to Cart