COM448685354 Combi-Blocks, Inc.

6-Fluoro-2-phenylbenzoic acid

  • MDL number MFCD14701904
  • Cas Number : 1841-56-1
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8656
Availability details for
Quantity
  • Availability
  • Add to Cart