COM448685361 Combi-Blocks, Inc.

2-Chloro-4-phenylbenzoic acid

  • MDL number MFCD14701682
  • Cas Number : 5728-40-5
  • Molecular Weight : 232.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8663

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8663
Availability details for
Quantity
  • Availability
  • Add to Cart