COM448685371 Combi-Blocks, Inc.

5-Chloro-3-(thiophen-2-yl)benzoic acid

  • MDL number MFCD18318866
  • Cas Number : 1261928-17-9
  • Molecular Weight : 238.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8726

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8726
Availability details for
Quantity
  • Availability
  • Add to Cart